SY3H3D67F449 Synthonix Corporation

7-hydroxy-3-(4-nitrophenyl)chromen-4-one

  • Cas Number : 15485-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58524.html

Safety Information

MSDS https://synthonix.com/58524.html
Availability details for
Quantity
  • Availability
  • Add to Cart