SY3H3D67F44F Synthonix Corporation

1-(4-Hydroxyphenyl)-2-(2-methoxyphenoxy)propane-1,3-diol

  • MDL number MFCD29047018
  • Cas Number : 56122-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58713.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58713.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart