SY3H3D67F450 Synthonix Corporation

(2R,3S,4R,5R)-2-(Hydroxymethyl)-5-(6-((2-(methylthio)ethyl)amino)-2-((3,3,3-trifluoropropyl)thio)-9H-purin-9-yl)tetrahydrofuran-3,4-diol

  • MDL number MFCD15832982
  • Cas Number : 163706-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58784.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58784.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart