SY3H3D67F455 Synthonix Corporation

3-Hydroxy-2-(2-methoxyphenoxy)-1-(4-methoxyphenyl)propan-1-one

  • MDL number MFCD28399425
  • Cas Number : 92409-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58971.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58971.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart