SY3H3D67F457 Synthonix Corporation

3-Hydroxy-N-(2-oxo-2,3-dihydro-1H-benzo[d]imidazol-5-yl)-2-naphthamide

  • MDL number MFCD00388332
  • Cas Number : 26848-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59157.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59157.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart