SY3H3D67F458 Synthonix Corporation

3-Hydroxy-N-(3-methoxyphenyl)-2-naphthamide

  • MDL number MFCD15542703
  • Cas Number : 62553-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59181.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59181.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart