SY3H3D67F45C Synthonix Corporation

10-Hydroxy-1-(2-hydroxy-3,4-dimethoxy-6-methylphenyl)decan-1-one

  • MDL number MFCD14584418
  • Cas Number : 104966-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59458.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59458.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart