SY3H3D67F45E Synthonix Corporation

4'-(Heptyloxy)-[1,1'-biphenyl]-4-carbonitrile

  • MDL number MFCD00134146
  • Cas Number : 52364-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59557.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59557.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart