SY3H3D67F460 Synthonix Corporation

3-Hexadecylthiophene

  • MDL number MFCD07368973
  • Cas Number : 119269-24-8
  • Molecular Weight : 308.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-hexadecylthiophene
Inchi Code 1S/C20H36S/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-20-17-18-21-19-20/h17-19H,2-16H2,1H3
Inchi Key FRVZSODZVJPMKO-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form Crystal - Lump / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/59601.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/59601.html
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart