SY3H3D67F46F Synthonix Corporation

5-Iodo-4-(trifluoromethyl)thiazol-2-amine

  • Cas Number : 682342-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/39985.html

Safety Information

MSDS https://synthonix.com/39985.html
Availability details for
Quantity
  • Availability
  • Add to Cart