SY3H3D67F470 Synthonix Corporation

4-Iodo-1H-pyrazole-5-carbonitrile

  • MDL number MFCD23135780
  • Cas Number : 1354703-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40007.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40007.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart