SY3H3D67F479 Synthonix Corporation

5-Iodo-3-methyl-1H-pyrazole

  • MDL number No MDL found
  • Cas Number : 1555820-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40122.html

Safety Information

MSDS https://synthonix.com/40122.html
Availability details for
Quantity
  • Availability
  • Add to Cart