SY3H3D67F47D Synthonix Corporation

5-Iodopyrimidine-2,4-diamine

  • MDL number No MDL found
  • Cas Number : 157924-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40153.html

Safety Information

MSDS https://synthonix.com/40153.html
Availability details for
Quantity
  • Availability
  • Add to Cart