SY3H3D67F484 Synthonix Corporation

(1H-Imidazol-4-yl)methanamine hydrochloride

  • Cas Number : 66247-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40332.html

Safety Information

MSDS https://synthonix.com/40332.html
Availability details for
Quantity
  • Availability
  • Add to Cart