SY3H3D67F487 Synthonix Corporation

3-Iodo-4-nitropyridine

  • MDL number No MDL found
  • Cas Number : 89283-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40591.html

Safety Information

MSDS https://synthonix.com/40591.html
Availability details for
Quantity
  • Availability
  • Add to Cart