SY3H3D67F491 Synthonix Corporation

3-Iodopyridin-2-ol

  • Cas Number : 1822890-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/40856.html

Safety Information

MSDS https://synthonix.com/40856.html
Availability details for
Quantity
  • Availability
  • Add to Cart