SY3H3D67F49A Synthonix Corporation

5-Iodo-4-methylpyrimidine

  • Cas Number : 91749-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40890.html

Safety Information

MSDS https://synthonix.com/40890.html
Availability details for
Quantity
  • Availability
  • Add to Cart