SY3H3D67F49F Synthonix Corporation

5-Iodo-2-methoxypyrimidine

  • MDL number No MDL found
  • Cas Number : 101803-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40945.html

Safety Information

MSDS https://synthonix.com/40945.html
Availability details for
Quantity
  • Availability
  • Add to Cart