SY3H3D67F4A1 Synthonix Corporation

5-Iodo-6-methylpyrimidin-4(1H)-one

  • Cas Number : 7752-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40947.html

Safety Information

MSDS https://synthonix.com/40947.html
Availability details for
Quantity
  • Availability
  • Add to Cart