SY3H3D67F4A2 Synthonix Corporation

4-Iodo-1-methyl-1H-pyrazole-5-carbaldehyde

  • MDL number No MDL found
  • Cas Number : 959986-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/40948.html

Safety Information

MSDS https://synthonix.com/40948.html
Availability details for
Quantity
  • Availability
  • Add to Cart