SY3H3D67F4A7 Synthonix Corporation

4-Iodo-1,3-dimethyl-1H-pyrazole

  • MDL number No MDL found
  • Cas Number : 6647-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41135.html

Safety Information

MSDS https://synthonix.com/41135.html
Availability details for
Quantity
  • Availability
  • Add to Cart