SY3H3D67F4AA Synthonix Corporation

2-(1H-Imidazol-1-yl)acetic acid hydrochloride

  • MDL number No MDL found
  • Cas Number : 87266-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41347.html

Safety Information

MSDS https://synthonix.com/41347.html
Availability details for
Quantity
  • Availability
  • Add to Cart