SY3H3D67F4AB Synthonix Corporation

2-(1H-Imidazol-5-yl)ethanol hydrochloride

  • MDL number No MDL found
  • Cas Number : 180307-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41362.html

Safety Information

MSDS https://synthonix.com/41362.html
Availability details for
Quantity
  • Availability
  • Add to Cart