SY3H3D67F4B3 Synthonix Corporation

4-Iodo-5-(trifluoromethyl)pyridin-2-ol

  • MDL number No MDL found
  • Cas Number : 1227601-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/42024.html

Safety Information

MSDS https://synthonix.com/42024.html
Availability details for
Quantity
  • Availability
  • Add to Cart