SY3H3D67F4B4 Synthonix Corporation

5-Iodo-6-(trifluoromethyl)pyridin-2-ol

  • MDL number No MDL found
  • Cas Number : 1227515-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/42025.html

Safety Information

MSDS https://synthonix.com/42025.html
Availability details for
Quantity
  • Availability
  • Add to Cart