SY3H3D67F4BC Synthonix Corporation

3-Iodopicolinic acid

  • MDL number No MDL found
  • Cas Number : 73841-32-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/42258.html

Safety Information

MSDS https://synthonix.com/42258.html
Availability details for
Quantity
  • Availability
  • Add to Cart