SY3H3D67F4BD Synthonix Corporation

3-Iodo-4-nitrophenol

  • MDL number No MDL found
  • Cas Number : 50590-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42284.html

Safety Information

MSDS https://synthonix.com/42284.html
Availability details for
Quantity
  • Availability
  • Add to Cart