SY3H3D67F4BE Synthonix Corporation

5-Iodo-2-nitrophenol

  • MDL number No MDL found
  • Cas Number : 27783-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/42285.html

Safety Information

MSDS https://synthonix.com/42285.html
Availability details for
Quantity
  • Availability
  • Add to Cart