SY3H3D67F4C1 Synthonix Corporation

Isoxazolo[5,4-c]pyridin-3-amine

  • MDL number No MDL found
  • Cas Number : 114080-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/42473.html

Safety Information

MSDS https://synthonix.com/42473.html
Availability details for
Quantity
  • Availability
  • Add to Cart