SY3H3D67F4C3 Synthonix Corporation

Imidazo[1,2-a]pyrimidine-5,7-diol

  • MDL number No MDL found
  • Cas Number : 51647-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42482.html

Safety Information

MSDS https://synthonix.com/42482.html
Availability details for
Quantity
  • Availability
  • Add to Cart