SY3H3D67F4C4 Synthonix Corporation

1H-Imidazo[4,5-b]pyridine-2(3H)-thione

  • MDL number No MDL found
  • Cas Number : 29448-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42494.html

Safety Information

MSDS https://synthonix.com/42494.html
Availability details for
Quantity
  • Availability
  • Add to Cart