SY3H3D67F4C6 Synthonix Corporation

2-Iodobenzenesulfonic acid

  • Cas Number : 63059-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/42719.html

Safety Information

MSDS https://synthonix.com/42719.html
Availability details for
Quantity
  • Availability
  • Add to Cart