SY3H3D67F4CA Synthonix Corporation

1H-Imidazo[4,5-b]pyridin-6-amine

  • MDL number No MDL found
  • Cas Number : 329946-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/42831.html

Safety Information

MSDS https://synthonix.com/42831.html
Availability details for
Quantity
  • Availability
  • Add to Cart