SY3H3D67F4D4 Synthonix Corporation

2-Iodo-1,4-benzenediamine

  • Cas Number : 69951-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43019.html

Safety Information

MSDS https://synthonix.com/43019.html
Availability details for
Quantity
  • Availability
  • Add to Cart