SY3H3D67F4D6 Synthonix Corporation

3-Iodo-2,5-dimethylpyrazine

  • Cas Number : 59021-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43021.html

Safety Information

MSDS https://synthonix.com/43021.html
Availability details for
Quantity
  • Availability
  • Add to Cart