SY3H3D67F4D7 Synthonix Corporation

2-Iodo-6-methylpyridin-3-amine

  • Cas Number : 1211596-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/43022.html

Safety Information

MSDS https://synthonix.com/43022.html
Availability details for
Quantity
  • Availability
  • Add to Cart