SY3H3D67F4D8 Synthonix Corporation

5-Iodo-2-methylpyridin-4-amine

  • MDL number No MDL found
  • Cas Number : 849353-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/43023.html

Safety Information

MSDS https://synthonix.com/43023.html
Availability details for
Quantity
  • Availability
  • Add to Cart