SY3H3D67F4DC Synthonix Corporation

(2-Iodophenyl)hydrazine hydrochloride

  • Cas Number : 60481-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/43028.html

Safety Information

MSDS https://synthonix.com/43028.html
Availability details for
Quantity
  • Availability
  • Add to Cart