SY3H3D67F4DD Synthonix Corporation

(4-Iodophenyl)hydrazine hydrochloride

  • MDL number No MDL found
  • Cas Number : 62830-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43029.html

Safety Information

MSDS https://synthonix.com/43029.html
Availability details for
Quantity
  • Availability
  • Add to Cart