SY3H3D67F4E0 Synthonix Corporation

Imidazo[1,2-a]pyrazin-8(7H)-one hydrobromide

  • MDL number No MDL found
  • Cas Number : 1451215-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43426.html

Safety Information

MSDS https://synthonix.com/43426.html
Availability details for
Quantity
  • Availability
  • Add to Cart