SY3H3D67F4E3 Synthonix Corporation

1-iodo-3-nitro-5-(trifluoromethyl)benzene

  • MDL number No MDL found
  • Cas Number : 41253-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44152.html

Safety Information

MSDS https://synthonix.com/44152.html
Availability details for
Quantity
  • Availability
  • Add to Cart