SY3H3D67F4E8 Synthonix Corporation

2-Iodobenzo[d]thiazole

  • Cas Number : 1123-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/44444.html

Safety Information

MSDS https://synthonix.com/44444.html
Availability details for
Quantity
  • Availability
  • Add to Cart