SY3H3D67F4E9 Synthonix Corporation

6-Iodobenzo[d]thiazole

  • MDL number No MDL found
  • Cas Number : 654070-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44445.html

Safety Information

MSDS https://synthonix.com/44445.html
Availability details for
Quantity
  • Availability
  • Add to Cart