SY3H3D67F4EC Synthonix Corporation

2-Iodo-5-nitrobenzoic acid

  • MDL number No MDL found
  • Cas Number : 19230-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/44515.html

Safety Information

MSDS https://synthonix.com/44515.html
Availability details for
Quantity
  • Availability
  • Add to Cart