SY3H3D67F4F4 Synthonix Corporation

6-Iodoimidazo[1,2-a]pyridine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1205744-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44807.html

Safety Information

MSDS https://synthonix.com/44807.html
Availability details for
Quantity
  • Availability
  • Add to Cart