SY3H3D67F4F8 Synthonix Corporation

Imidazo[1,2-a]pyrimidine-6-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 944896-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/44880.html

Safety Information

MSDS https://synthonix.com/44880.html
Availability details for
Quantity
  • Availability
  • Add to Cart