SY3H3D67F502 Synthonix Corporation

Imidazo[1,2-a]pyridin-8-ol

  • MDL number No MDL found
  • Cas Number : 69214-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45218.html

Safety Information

MSDS https://synthonix.com/45218.html
Availability details for
Quantity
  • Availability
  • Add to Cart