SY3H3D67F50B Synthonix Corporation

2-Iodo-1-methyl-4-nitrobenzene

  • MDL number No MDL found
  • Cas Number : 7745-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/45401.html

Safety Information

MSDS https://synthonix.com/45401.html
Availability details for
Quantity
  • Availability
  • Add to Cart