SY3H3D67F50C Synthonix Corporation

4-Iodo-2-methoxynicotinaldehyde

  • MDL number No MDL found
  • Cas Number : 158669-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/45402.html

Safety Information

MSDS https://synthonix.com/45402.html
Availability details for
Quantity
  • Availability
  • Add to Cart