SY3H3D67F50F Synthonix Corporation

1-Iodo-4-methoxy-2-nitrobenzene

  • MDL number MFCD00024195
  • Cas Number : 58755-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Light-yellow to yellow to brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45456.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45456.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart